4-Allylveratrole

Supplier: Roth Carl
Warning

Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol

5073.1
ROTH5073.1EA 0 EUR
ROTH5073.1
4-Allylveratrole
4-Allylveratrole
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 255
Melting Pt: –4 °C
Density: 1.036 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008652
CAS Number: 93-15-2
EINECS: 202-223-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR