Anethole

Supplier: Roth Carl

5131.1 5131.2
ROTH5131.1EA 0 EUR
ROTH5131.1 ROTH5131.2
Anethole
Anethole
Formula: C₁₀H₁₂O
Boiling Pt: 234,5 °C (1013 hPa)
Melting Pt: 21,3 °C
Density: 1 g/cm³ (12 °C)
MDL Number: MFCD00009284
CAS Number: 104-46-1
EINECS: 203-205-5
Merck Index: 13,00647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR