Carboxymethyl cellulose sodium salt

Supplier: Roth Carl

6190.1 6190.2
ROTH6190.1EA 0 EUR
ROTH6190.1 ROTH6190.2
Carboxymethyl cellulose sodium salt
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR