5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

Supplier: Roth Carl
Warning

6334.1
ROTH6334.1EA 0 EUR
ROTH6334.1
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Formula: C₁₄H₈N₂O₈S₂
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR