Cyclohexane

Supplier: Roth Carl
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

6570.1 6570.2 6570.3 6570.4
ROTH6570.1EA 0 EUR
ROTH6570.1 ROTH6570.2 ROTH6570.3 ROTH6570.4
Cyclohexane
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR