tert-Butyl methyl ether

Supplier: Roth Carl
Danger

6746.1 6746.2 6746.3 6746.4
ROTH6746.1EA 0 EUR
ROTH6746.1 ROTH6746.2 ROTH6746.3 ROTH6746.4
tert-Butyl methyl ether
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
Boiling Pt: 55,2 °C (1013 hPa)
Melting Pt: –108,6 °C
Density: 0,7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR