TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)

Supplier: Roth Carl

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

6982.1 6982.2 6982.3 6982.4
ROTH6982.1EA 0 EUR
ROTH6982.1 ROTH6982.2 ROTH6982.3 ROTH6982.4
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS Number: 29915-38-6
EINECS: 249-954-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR