(S)-(-)-Nicotine

Supplier: Roth Carl
Danger

8746.1
ROTH8746.1EA 0 EUR
ROTH8746.1
(S)-(-)-Nicotine
(S)-(-)-Nicotine
Formula: C₁₀H₁₄N₂
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: –79 °C
Density: 1,01 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1,II
Merck Index: 12,06611

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR