PIPES

Supplier: Roth Carl

9156.1 9156.2 9156.3 9156.4
ROTH9156.1EA 0 EUR
ROTH9156.1 ROTH9156.2 ROTH9156.3 ROTH9156.4
PIPES
PIPES
Formula: C₈H₁₈N₂O₆S₂
Boiling Pt: 499 °C (1013 hPa)
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
Merck Index: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR