Veratraldehyde

Supplier: Roth Carl
Warning

9651.1 9651.2
ROTH9651.1EA 0 EUR
ROTH9651.1 ROTH9651.2
Veratraldehyde
Veratraldehyde
Formula: C₉H₁₀O₃
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 40…43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR