4-Hydroxycinnamic acid

Supplier: Roth Carl
Warning

9906.1
ROTH9906.1EA 0 EUR
ROTH9906.1
4-Hydroxycinnamic acid
4-Hydroxycinnamic acid
Formula: C₉H₈O₃
MDL Number: MFCD00004399
CAS Number: 7400-08-0
EINECS: 231-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR