Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution

Supplier: Roth Carl
Warning

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

P719.1
ROTHP719.1EA 116 EUR
ROTHP719.1
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8 III
Merck Index: 12,02036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR