Methyl orange

Supplier: Roth Carl
Danger

T118.1 T118.2
ROTHT118.1EA 45.1 EUR
ROTHT118.1 ROTHT118.2
Methyl orange
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: > 300 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 13,06127

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR