Acetonitrile

Supplier: Roth Carl
Danger

T195.1 T195.2
ROTHT195.1EA 139 EUR
ROTHT195.1 ROTHT195.2
Acetonitrile
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR