Thiamine hydrochloride

Supplier: Roth Carl

T911.1 T911.2
ROTHT911.1EA 35.5 EUR
ROTHT911.1 ROTHT911.2
Thiamine hydrochloride
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
Melting Pt: 252 °C
Density: 1,401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR