Methyl ethyl ketone

Supplier: Roth Carl
Danger

T920.1 T920.2
ROTHT920.1EA 64.4 EUR
ROTHT920.1 ROTHT920.2
Methyl ethyl ketone
Methyl ethyl ketone
Formula: C₂H₅COCH₃
Boiling Pt: 79,6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0,8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR