Sodium sulphate decahydrate

Supplier: Roth Carl

X892.1 X892.2 X892.3
ROTHX892.1EA 28.3 EUR
ROTHX892.1 ROTHX892.2 ROTHX892.3
Sodium sulphate decahydrate
Sodium sulphate decahydrate
Formula: Na₂SO₄·10H₂O
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 32,4 °C
Density: 1,464 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3
EINECS: 231-820-9
Merck Index: 13,08755

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR