Safety storage cabinets for flammable liquids
Supplier: SAFETY UNLIMITED
SC1010 SC1030 SC1040 SC1050 SC1060 SC1080
SAFUSC1010EA 654 EUR
SAFUSC1010 SAFUSC1030 SAFUSC1040 SAFUSC1050 SAFUSC1060 SAFUSC1080
Safety storage cabinets for flammable liquids
Furniture Storage Cabinets
For the safety storage of flammable liquids. Models with single and double doors with fully adjustable shelves to 45 mm pitch are available. Doors have a reinforced vertical panel to prevent distortion due to heat. Shelves are perforated to allow spillages to drain into the leakproof sump below, with sump cover to allow containers to be stored on the sump while retaining the full containment capacity.
  • Spot welded seams with gas welded corner joints
  • Lockable via a two point locking system
  • Removable leakproof sump
Made from zintec coated steel, finishedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable