Transfer pipettes, general purpose, Samco™
Supplier: SAMCO
202-1S 202-20S 202 273 273-1S 336-1S 335-11
SAMC202-1SEA 0 EUR
SAMC202-1S SAMC202-20S SAMC202 SAMC273 SAMC273-1S SAMC336-1S SAMC335-11
Transfer pipettes, general purpose, Samco™
Pipettes Pasteur/Transfer Pipettes
These transfer pipettes are made of shatterproof, non toxic polyethylene. They have a built-in bulb that eliminates the need for plugging, packaging and autoclaving.
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Pipettes can be frozen in liquid nitrogen, or heat sealed for storage
  • Non graduated
Ideal for tissue culture applications, routing sampling of serum and plasma, blood culture, urine analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)