Inserts for U-D master manual samplers
Supplier: Sampling Systems
1230T-05S 1230T-10S 1230T-30S 1230T-03S 1230T-15S
SAMP1230T-05SEA 394 EUR
SAMP1230T-05S SAMP1230T-10S SAMP1230T-30S SAMP1230T-03S SAMP1230T-15S
Inserts for U-D master manual samplers
Samplers Solid Samplers
Stainless steel inserts for U-D Master manual samplers.
  • Made from 316 stainless steel
These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.
Order Now
SPECIFICATIONS
Environmentally Preferable