Enviro Dipper Sets
Supplier: Sampling Systems
5085A-01
SAMP5085A-01EA 105 EUR
SAMP5085A-01
Enviro Dipper Sets
Samplers Sample Dippers
The enviro dipper is great for the environmental sampling of liquids.
  • Includes 300 ml stainless steel bucket, hand reel and 50 m of PP rope
  • Hand reel incorporates a built in ratchet mechanism and brake
The enviro dipper is a complete set with everything you need to enable the fast and efficient environmental sampling of liquids across a wide variety of applications including from streams and rivers. It is ideal for contamination monitoring, regulatory compliance and research studies.