Filtration capsules, Sartoclean® GF MidiCaps
Supplier: Sartorius
5605304E9-OO-A
SARP5605304E9-OO-AEA 914 EUR
SARP5605304E9-OO-A
Filtration capsules, Sartoclean® GF MidiCaps
Filters Membrane Filter Capsules
Ready-to-connect units contains a multi-step combination of filters for effective and economical particle removal.
  • Ultra-low unspecific binding membrane
  • More economical process flow
  • Increased total throughput performance
The heterogeneous double layer is protected by a layer of glass fleece. This increased the total throughput performance of the membrane double layer and adds an adsorptive component to filtration process.
Order Now
SPECIFICATIONS
Environmentally Preferable
Inlet/outlet OO (Hose Barb/Hose Barb)
Ø×L 77×197 mm
Max. pressure 5 bar @ 20 °C