Transport Cooling Containers
Supplier: SARSTEDT
95.1011 95.1123
SARS95.1011EA 109 EUR
SARS95.1011 SARS95.1123
Transport Cooling Containers
Boxes Cooling/Freezing Boxes and Bags
The cool transport containers is suitable for safe cool transportation of medical and biological specimens. It can accommodate two sample tubes of 10 ml volume or any tubes of 16,5 mm Ø and 107 mm length.
The mailing container is filled with a coolant and should be frozen in a horizontal position at −12 °C before mailing. The styrofoam casing protects against warmth and any damage during transport. The container is not suitable for samples that are not allowed to freeze.