Tork Surface Disinfectant Sprays
Supplier: Essity
193610
SCAA193610EA 29.5 EUR
SCAA193610
Tork Surface Disinfectant Sprays
Cleaning Agents Disinfectants
The surface disinfecting spray is suitable for PT2 (disinfectants not intended for direct application to humans or animals) and PT4 (food and feed area) applications, food contact surfaces must be rinsed with drinking water after disinfecting with product. The product is an alcohol-free ready-to-use multi-purpose spray for surface disinfection.
  • Kills 99,99% of enveloped viruses in five minutes including coronavirus EN14476: Limited virucidal activity (Active against enveloped viruses such as, e.g. Influenza virus, coronavirus, HIV, herpes virus, hepatitis
More Product Information