Insulated shippers, AmbiTech®, ThermoSafe®
Supplier: Sonoco Thermosafe
ATR18 ATG4 ATR34 ATR71 ATG9 ATG17 ATG20 ATG43
SCATATR18EA 4130 EUR
SCATATR18 SCATATG4 SCATATR34 SCATATR71 SCATATG9 SCATATG17 SCATATG20 SCATATG43
Insulated shippers, AmbiTech®, ThermoSafe®
Mailers and Shipping Containers
These moulded containers are designed for logistics applications in the pharmaceutical industries, where temperature-sensitive products have to be transported and stored in a temperature range of 15 to 25 °C.
  • Simple and quick assembly with minimal components, saves time
  • Provides excellent stability in extreme ambient conditions over extended durations
Regional range containers provides protection for up to 72 hours whereas global range provides 120 hours.
Certifications: Tested to ISTA 7D and 7E temperature profiles.
Order Now
SPECIFICATIONS
Environmentally Preferable