ThermoSafe® Certis® Pre-Qualified 2 to 8 °C Shippers
Supplier: Sonoco Thermosafe
C26R5-13
SCATC26R5-13EA 0 EUR
SCATC26R5-13
ThermoSafe® Certis® Pre-Qualified 2 to 8 °C Shippers
Mailers and Shipping Containers
Certis® shippers are designed so that there is no need for bench conditioning of the refrigerants, saving time space and money.
  • Pre-qualified 2 to 8 °C shipper delivering 5 day or 120 hour performance
  • Includes VIP insulation panels and PCM refrigerants to provide an exceptional level of temperature control
  • Qualified duration: 5 days
  • Low volumetric weight saves shipping costs
Certis® is a family of premium, universal shippers for extended durations. Universal solutions designed to be able to ship across time zones and climates. Certis® 5 DayMore Product Information