Microfiltration assembly, with fritted glass support, ULTRA-WARE®, KIMBLE®
Supplier: DWK Life Sciences
SP953705-0000 SP953755-0000 SP953755-0090 SP953780-0000
SCERSP953705-0000EA 806 EUR
SCERSP953705-0000 SCERSP953755-0000 SCERSP953755-0090 SCERSP953780-0000
Microfiltration assembly, with fritted glass support, ULTRA-WARE®, KIMBLE®
Filters Membrane Filter Units
These vacuum microfiltration assemblies are manufactured from 33 expansion, low extractable borosilicate glass. The 47 mm vacuum microfiltration assembly is used for general filtration and is designed to handle up to 500 ml of sample liquids for the analysis of particulate or microbiological contamination. The 90 mm vacuum microfiltration assembly is designed for large sample volumes or samples with high particulate loads that would tend to clog a 47 mm diameter filter. The assembly with fritted glass support is used for general filtration.
  • Funnel is graduated from 100 to 250 ml in 25
More Product Information