Thermometer for pycnometer
Supplier: Ludwig Schneider GmbH & CO. KG
1322107S
SCNL1322107SEA 260 EUR
SCNL1322107S
Thermometer for pycnometer
Thermometers Analogue Thermometers
Thermometer for pycnometer in standard ground joint size NS 10/19, enclosed type.
  • Colourless prismatic capillary
  • ACCU-SAFE filling
  • Total immersion depth
For 25 ml content. Suitable for official certification.
Certifications: According to DIN 12809.