DURAN® Sealing Rings, for Polished Sperical Joints (VITON)
Supplier: DWK Life Sciences
1155355 1155356
SCOT1155355EA 49.2 EUR
SCOT1155355 SCOT1155356
DURAN® Sealing Rings, for Polished Sperical Joints (VITON)
Seals Sealing Rings
Ball joints without ground joints only seal via the sealing ring.
  • Non autoclavable
  • Used in vacuum applications
Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.