DURAN® Flasks, Round Bottom, with Two Necks and Standard Gound Joints
Supplier: DWK Life Sciences
241832604 241832707 241833609 241834408 241835404 241833703 241835601
SCOT241832604EA 56.2 EUR
SCOT241832604 SCOT241832707 SCOT241833609 SCOT241834408 SCOT241835404 SCOT241833703 SCOT241835601
DURAN® Flasks, Round Bottom, with Two Necks and Standard Gound Joints
Flasks Round Bottom Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one parallel side neck. With matt white labelling area.
*non-DIN ISO size.
Certifications: DIN 12394.
Order Now
SPECIFICATIONS
Environmentally Preferable