Gas washing bottles, DURAN®
Supplier: DWK Life Sciences
257030109
SCOT257030109EA 326 EUR
SCOT257030109
Gas washing bottles, DURAN®
Bottles Gas Washing Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sealed-in fritted dis, with standard ground joint and cap.