Stopcock, single way, taper 1:5, with PTFE key, DURAN®
Supplier: DWK Life Sciences
286453009 286453806 286453206 286453403 286454005 286454202
SCOT286453009EA 1320 EUR
SCOT286453009 SCOT286453806 SCOT286453206 SCOT286453403 SCOT286454005 SCOT286454202
Stopcock, single way, taper 1:5, with PTFE key, DURAN®
Stopcocks
DURAN®, borosilicate glass 3.3, clear, with PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.