Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
317010 397010 397050 397008 397100
SCTV317010EA 35.6 EUR
SCTV317010 SCTV397010 SCTV397050 SCTV397008 SCTV397100
Air valves for safetycaps generation 2.0
Valves Air Regulation Valves
These air valves can be used for analytical and preperative applications.
  • PTFE filter for clean solvents
  • Valve membrane blocks solvent vapours, opens at underpressure
  • Thread size UNF 1/4" 28G
They include a luer lock adapter which is useful for many applications.
Order Now
SPECIFICATIONS
Environmentally Preferable