Boiling baths, Precisterm
Supplier: SELECTA
6000386
SELE6000386EA 0 EUR
SELE6000386
Boiling baths, Precisterm
Baths Boiling Baths
Double skinned AISI 310 stainless steel tank with an exterior case of AISI 304 stainless steel. The heating elements embedded in INCOLOY heat and corrosion resistant stainless steel.
  • Illuminated mains power switch
  • Hydraulic thermostat temperature control, synchronised with the 'ON' indicator lamp
  • Safety thermostat cut out indicator lamp (DIN 12877.2)