Tubing, silicone, platinum cured

Supplier: SF MEDICAL
SFM3-2050 SFM3-4250
SFAASFM3-2050EA 277 EUR
SFAASFM3-2050 228-0260
Tubing, silicone, platinum cured
Tubing Transfer Tubing
Silicone, platinum cured.

  • Extended post cure for lowest level extractables
  • Biocompatible, inert, non-reactive with other elements per systemic testing
  • Odourless, tasteless, non-toxic for use in medical, food, drug, personal care, and deionised water applications
  • Non-leaching plasticisers
  • Sterilizable by radiation, ETO, steam –30 bar or mbar at 253 °F (123 °C) for use in sterile applications, reusability
  • Colour stripe, tint for custom process control identification

Temperature resistance:—54 to +204 °C

Hardness (Shore durometer A): 60

Density: 1,15
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR