HyClone™ Fetal Bovine Serum Characterized (Australia), HyClone Products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30084.03IR25 SH30084.03IR30
SH30084.03IR25EA 1300 EUR
SH30084.03IR25 SH30084.03IR30
HyClone™ Fetal Bovine Serum Characterized (Australia), HyClone Products (Cytiva)
Sera (Serum)
This premium HyCloneTM Fetal Bovine Australian-origin Serum is well-characterized, triple 100 nm filtered, virus-tested to 9 CFR standard, and traceability-certified (ISIA, Oritain).
  • Benefit from Australian husbandry methods and certified traceability
  • Assayed for a variety of critical components and biochemical characteristics
  • Certificate of analysis confirms ≤25 EU/ml endotoxin
  • Serial 100 nm pore size-rated filtering for reliable microbial depletion
  • Gamma rradiated and heat-inactivated FBS options also available
  • Broad virus panel testing in accordance
More Product Information