Human Recombinant GACRP-30 (from E. coli)
Supplier: Shenandoah Biotechnology
100-156AF-5UG 100-156AF-25UG 100-156AF-100U 100-156AF-500U 100-156AF-1MG
SHBT100-156AF-5UGEA 78.1 EUR
SHBT100-156AF-5UG SHBT100-156AF-25UG SHBT100-156AF-100U SHBT100-156AF-500U SHBT100-156AF-1MG
Human Recombinant GACRP-30 (from E. coli)
Proteins and Peptides
The globular subunit of adipocyte complement-related protein of 30 kDa (ACRP-30) is a naturally occurring cleavage product of adiponectin, a protein made exclusively by adipocytes. ACRP-30 is an abundant serum protein and plays an important role in hyperglycemia, insulin resistance, and fatty acid oxidation. ACRP-30 signals through adiponectin receptor 1 (AdipoR1) and adiponectin receptor 2 (AdipoR2).
  • High quality
  • Low endotoxin
  • Affordable
Caution: This product is for research use only and is not for use in humans.