Human Recombinant FGF-10 (from E. coli)
Supplier: Shenandoah Biotechnology
100-183-100UG 100-183-1MG 100-183-25UG 100-183-500UG 100-183-5UG
SHBT100-183-100UGEA 453 EUR
SHBT100-183-100UG SHBT100-183-1MG SHBT100-183-25UG SHBT100-183-500UG SHBT100-183-5UG
Human Recombinant FGF-10 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis.FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinising epidermal cells, and induces the migration and invasion of cancer cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.