Mouse Recombinant IL-27/P28 (from E. coli)

Supplier: Shenandoah Biotechnology

200-50AF-100UG 200-50AF-10UG 200-50AF-1MG 200-50AF-500UG
SHBT200-50AF-100UGEA 766 EUR
SHBT200-50AF-100UG SHBT200-50AF-10UG SHBT200-50AF-1MG SHBT200-50AF-500UG
Mouse Recombinant IL-27/P28 (from E. coli)
Proteins and Peptides

The p28 subunit of interleukin 27 (IL-27), also known as interleukin 30 (IL-30), is a member of the interleukin 12 (IL-12) family of cytokines. p28 is a secreted polypeptide that associates with the Epstein-Barr virus induced gene 3 (EBI3) to form the IL-27 cytokine heterodimer complex. IL-27 functions as a proinflammatory cytokine that induces immunomodulatory effects in naïve CD4+ T cells, mast cells, and monocytes. p28 can also form a complex with cytokine-like factor 1 (CLF), that is secreted by dendritic cells, to regulate natural killer (NK) and T cell functions.


  • High quality
  • Low endotoxin
  • Affordable


Caution: This product is for research use only and is not for use in humans.

Order Now

SPECIFICATIONS

Protein/peptide name IL-27/P28
Environmentally Preferable
Protein synonyms P28|IL-27 p28 subunit|IL-30
Protein/peptide type Recombinant
Protein function Cytokine
Species Mouse
Source E. coli
Conjugation Unconjugated
Gene ID Q8K3I6
Sequence MFPTDPLSLQ ELRREFTVSL YLARKLLSEV QGYVHSFAES RLPGVNLDLL PLGYHLPNVS LTFQAWHHLS DSERLCFLAT TLRPFPAMLG GLGTQGTWTS SEREQLWAMR LDLRDLHRHL RFQVLAAGFK CSKEEEDKEE EEEEEEEEKK LPLGALGGPN QVSSQVSWPQ LLYTYQLLHS LELVLSRAVR DLLLLSLPRR PGSAWDS
Protease-free N
Animal-free Y
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤1
Biological activity Bioactivity: No biological activity data is available at this time
Purity ≥95%
Grade RUO
Formulation 10 mM Sodium bicarbonate, pH 8,5
Molecular weight 23,7 kDa
Reconstitution instructions Sterile water at 0,1 mg/ml
Storage conditions –20 °C
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR