Mouse Recombinant IL-17E (from E. coli)
Supplier: Shenandoah Biotechnology
200-60-100UG 200-60-1MG 200-60-25UG 200-60-500UG 200-60-5UG
SHBT200-60-100UGEA 453 EUR
SHBT200-60-100UG SHBT200-60-1MG SHBT200-60-25UG SHBT200-60-500UG SHBT200-60-5UG
Mouse Recombinant IL-17E (from E. coli)
Proteins and Peptides
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.