Mouse Recombinant IL-10 (from E. coli)
Supplier: Shenandoah Biotechnology
200-84AF-10UG 200-84AF-100UG 200-84AF-500UG 200-84AF-1MG
SHBT200-84AF-10UGEA 199 EUR
SHBT200-84AF-10UG SHBT200-84AF-100UG SHBT200-84AF-500UG SHBT200-84AF-1MG
Mouse Recombinant IL-10 (from E. coli)
Proteins and Peptides
Interleukin 10 (IL-10) is an anti-inflammatory cytokine produced by macrophages and type 2 T helper (Th2) cells. IL-10 inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFN-ƴ), tumor necrosis factor alpha (TNF-α), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. IL-10 also suppresses antigen presentation on antigen presenting cells, and enhances the survival, proliferation, and antibody production of B cells. Mouse IL-10 is not active on human cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: This product is for research use only and is not for use in humans.