Rat Recombinant VEGF-165 (from E. coli)
Supplier: Shenandoah Biotechnology
300-31-100UG 300-31-10UG 300-31-1MG 300-31-500UG
SHBT300-31-100UGEA 525 EUR
SHBT300-31-100UG SHBT300-31-10UG SHBT300-31-1MG SHBT300-31-500UG
Rat Recombinant VEGF-165 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
These research grade recombinant proteins are typically used as cytokines, growth factors and chemokines in cell based assays and cell culture.
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.