Klerwipe™ Sporicide Wipes, Low Residue Peroxide Blended with WFI
Supplier: ECOLAB CONTAMINATION CONTROL
6600588
SHIE6600588EA 1050 EUR
SHIE6600588
Klerwipe™ Sporicide Wipes, Low Residue Peroxide Blended with WFI
Wipes Disposable Wipes and Towels Disinfectant Wipes
Sterile 100% polyester wipes impregnated with a very low residue blend of 6% hydrogen peroxide.
  • Ready-to-use
  • Very low residue rotational sporicide wipe
  • For small surfaces
The biocide blend is 0,2 µm filtered and sterile filled under Grade A laminar flow and double bagged in a Grade B cleanroom.
Packaging: Packed in a resealable pouch.