Organic size exclusion chromatography columns, GPC K-800 series preparative type
Supplier: Resonac
F6102306 F6102309 F6102312 F6102315 F6102304 F6102303 F6102305 F6102301 F6700407
SHOWF6102306EA 5011 EUR
SHOWF6102306 SHOWF6102309 SHOWF6102312 SHOWF6102315 SHOWF6102304 SHOWF6102303 SHOWF6102305 SHOWF6102301 SHOWF6700407
Organic size exclusion chromatography columns, GPC K-800 series preparative type
Chromatography Columns
These are styrene divinylbenzene copolymer-based organic solvent SEC (GPC) columns. Suitable for a wide range of applications from low to high molecular weight compounds.