Aqueous size exclusion chromatography columns, OHpak SB-800 HQ series preparative type
Supplier: Resonac
F6709555 F6516017 F6516011 F6516012 F6516013 F6516014 F6516016 F6516015
SHOWF6709555EA 844 EUR
SHOWF6709555 SHOWF6516017 SHOWF6516011 SHOWF6516012 SHOWF6516013 SHOWF6516014 SHOWF6516016 SHOWF6516015
Aqueous size exclusion chromatography columns, OHpak SB-800 HQ series preparative type
Chromatography Columns
SB-800 HQ are polyhydroxymethacrylate-based SEC columns for the molecular weight distribution analysis of water-soluble polymers. The solvent can be substituted with DMF (except for SB-802 HQ and SB-807 HQ columns), enabling the analysis of polar polymers.
  • Polymer-based SEC (GFC) columns for aqueous systems
  • Support a wide range of molecular weight
The maximum solvent tolerance of the SB-2000 series for methanol, acetonitrile and DMF is 50%. Solvent tolerance differs from that of the standard SB-800 HQ columns.