Ultrasonic wall thickness gauges, PosiTector®
Supplier: Industrial Physics
LD7025 LD7030 LD7108 LD7105 LD7107 LD7109 LD7110 LD7106
SIMELD7025EA 25.66 EUR
SIMELD7025 SIMELD7030 SIMELD7108 SIMELD7105 SIMELD7107 SIMELD7109 SIMELD7110 SIMELD7106
Ultrasonic wall thickness gauges, PosiTector®
Coating Testers
The PosiTector can measure the wall thickness of materials (plastic, stell) using ultrasonic technology. These durable and weatherproof models are resistant to solvents, acids, oils, water and dusts.
  • Precision transducers for fast and accurate readings
  • Built-in temperature compensation for higher accuracy
  • Transfer data via USB to a PC/Mac, Bluetooth wireless technology, Wi-Fi
Certifications: Traceable to NIST.
Delivery information: Supplied with probe, ultrasonic gel, rubber holster, belt clip, wrist strap, three batteries, instructions manual,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Batteries 3× AAA
Display High contrast reversible colour LCD
W×D×H 146×61×28 mm
Weight 140 g