Sample tubes for Roche Cobas analysers
Supplier: Simport Scientific
B721-1
SIMPB721-1EA 393 EUR
SIMPB721-1
Sample tubes for Roche Cobas analysers
Tubes Test Tubes
PS, clear.
  • Good chemical resistance
  • Good temperature resistance
This tube is used with the Roche Cobas Analyzer. It has perfect clarity for easy viewing of contents. This graduated false bottom tube has a capacity of 2,5 ml and will accept bar code labels.