Storage Boxes for Tubes

Supplier: Simport Scientific
T330-25B T330-25G T330-25P T330-25Y
SIMPT330-25BEA 292 EUR
SIMPT330-25B SIMPT330-25G SIMPT330-25P SIMPT330-25Y
Storage Boxes for Tubes
Boxes Boxes for Tubes
Cover is made of PS and base made of high impact PS.

  • Stackable
  • Colour coded grid
  • Air vents minimising condensation

These 5 ml tube storage boxes are designed to store samples between –90 to +80 °C.

Printed with a series of numbered squares and the surface accepts the writing with markers by facilitating inventory control.

Transparent cover that allows the user to see the contents of the box and is keyed to the base to prevent misalignment.
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR