Food processing sleeve protectors, Cork 8161
Supplier: SIOEN
8161Z0FC1A41 8161A2FK0W02
SIOE8161Z0FC1A41EA 16.2 EUR
SIOE8161Z0FC1A41 SIOE8161A2FK0W02
Food processing sleeve protectors, Cork 8161
Workwear
These waterproof sleeve protectors are made of Flexothane® Kleen fabric, a polyamide knitting 6.6 with PU coating.
  • Washable at 95 °C
  • Elasticated at both ends
  • High frequency welded seams
Ideal for food handling industries, green spaces and agriculture.
Weight: 170 g/m²