Bottle top vacuum filtration systems, Millicup™
Supplier: Merck
SJHVM4710
SJHVM4710EA 198 EUR
SJHVM4710
Bottle top vacuum filtration systems, Millicup™
Filters Bottle Top Filters
Millicup™ membrane filter units are non sterile universal bottle top vacuum filters, used for sterile filtration.
Max. temperature: 45 °C